SY3H3D67E5E3 Synthonix Corporation

1-(5-Fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine

  • Cas Number : 343-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54609.html

Safety Information

MSDS https://synthonix.com/54609.html
Availability details for
Quantity
  • Availability
  • Add to Cart