SY3H3D67E5EB Synthonix Corporation

2-(2-Fluorophenyl)pyrimidine-5-carbaldehyde

  • MDL number MFCD09743685
  • Cas Number : 946707-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Off white to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55190.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/55190.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart