SY3H3D67E5F7 Synthonix Corporation

(R)-2-(5-Fluoro-2-methoxyphenyl)pyrrolidine hydrochloride

  • Cas Number : 1260845-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55480.html

Safety Information

MSDS https://synthonix.com/55480.html
Availability details for
Quantity
  • Availability
  • Add to Cart