SY3H3D67E5FB Synthonix Corporation

5-(3-Fluorophenyl)-2-methylpyridine

  • MDL number MFCD13152238
  • Cas Number : 713143-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55577.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55577.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart