SY3H3D67E5FD Synthonix Corporation

4-(5-Formylthiophen-2-yl)-2-methylphenol

  • MDL number MFCD18313179
  • Cas Number : 1261976-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55616.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55616.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart