SY3H3D67E612 Synthonix Corporation

6-(4-Fluorophenyl)pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD28405285
  • Cas Number : 1036762-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to gray to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56429.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56429.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart