SY3H3D67E613 Synthonix Corporation

5-(2-Fluorophenyl)nicotinic acid

  • Cas Number : 705961-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56449.html

Safety Information

MSDS https://synthonix.com/56449.html
Availability details for
Quantity
  • Availability
  • Add to Cart