SY3H3D67E61B Synthonix Corporation

6-(3-Fluorophenyl)nicotinaldehyde

  • MDL number MFCD04115444
  • Cas Number : 898795-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56469.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56469.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart