SY3H3D67E61F Synthonix Corporation

6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride

  • MDL number MFCD06658529
  • Cas Number : 84163-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56574.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56574.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart