SY3H3D67E625 Synthonix Corporation

4-[[(4-Fluorophenyl)imino]methyl]phenol

  • Cas Number : 3382-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56675.html

Safety Information

MSDS https://synthonix.com/56675.html
Availability details for
Quantity
  • Availability
  • Add to Cart