SY3H3D67E630 Synthonix Corporation

(3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine

  • MDL number MFCD06658161
  • Cas Number : 105812-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57109.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57109.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart