SY3H3D67E634 Synthonix Corporation

6-Fluoro-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • Cas Number : 1227210-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57162.html

Safety Information

MSDS https://synthonix.com/57162.html
Availability details for
Quantity
  • Availability
  • Add to Cart