SY3H3D67E635 Synthonix Corporation

(3-fluoro-4-(3-morpholinopropoxy)phenyl)boronic acid

  • MDL number MFCD22418293
  • Cas Number : 944279-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57175.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57175.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart