SY3H3D67E63E Synthonix Corporation

2-((4-Fluorophenyl)ethynyl)aniline

  • MDL number MFCD00168851
  • Cas Number : 1173153-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57483.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57483.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart