SY3H3D67E643 Synthonix Corporation

3-(4-Fluorophenyl)phenethyl alcohol

  • Cas Number : 840521-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57631.html

Safety Information

MSDS https://synthonix.com/57631.html
Availability details for
Quantity
  • Availability
  • Add to Cart