SY3H3D67E647 Synthonix Corporation

2-(4-(2-Fluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD08706126
  • Cas Number : 864754-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57911.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57911.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart