SY3H3D67E64B Synthonix Corporation

1-(4-Fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile

  • MDL number MFCD06200812
  • Cas Number : 64169-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58121.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58121.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart