SY3H3D67E651 Synthonix Corporation

1-(2'-Fluoro-[1,1'-biphenyl]-4-yl)propan-1-one

  • Cas Number : 37989-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58213.html

Safety Information

MSDS https://synthonix.com/58213.html
Availability details for
Quantity
  • Availability
  • Add to Cart