SY3H3D67E653 Synthonix Corporation

(3-Fluoro-4'-propyl-[1,1'-biphenyl]-4-yl)boronic acid

  • Cas Number : 909709-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58281.html

Safety Information

MSDS https://synthonix.com/58281.html
Availability details for
Quantity
  • Availability
  • Add to Cart