SY3H3D67E654 Synthonix Corporation

(R)-(3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl)methyl methanesulfonate

  • MDL number MFCD18070719
  • Cas Number : 174649-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58337.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58337.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart