SY3H3D67E655 Synthonix Corporation

N-(2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propane-1-sulfonamide

  • MDL number MFCD22494591
  • Cas Number : 1269233-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58461.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58461.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart