SY3H3D67E65C Synthonix Corporation

4-Fluoro-2-(p-tolylethynyl)benzaldehyde

  • Cas Number : 1322091-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58581.html

Safety Information

MSDS https://synthonix.com/58581.html
Availability details for
Quantity
  • Availability
  • Add to Cart