SY3H3D67E666 Synthonix Corporation

4-(3-Fluoro-4-nitrophenoxy)-6,7-dimethoxyquinoline hydrochloride

  • MDL number MFCD31692616
  • Cas Number : 2232877-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59101.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59101.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart