SY3H3D67E66C Synthonix Corporation

(S)-(9H-Fluoren-9-yl)methyl (1-oxopropan-2-yl)carbamate

  • MDL number MFCD01318541
  • Cas Number : 146803-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59200.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59200.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart