SY3H3D67E677 Synthonix Corporation

4-(3-Fluorophenyl)-2,2-dimethyl-5-(4-(methylthio)phenyl)furan-3(2H)-one

  • MDL number MFCD22570932
  • Cas Number : 301693-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59383.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59383.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart