SY3H3D67E67C Synthonix Corporation

(2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid

  • MDL number MFCD00153365
  • Cas Number : 157355-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59404.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59404.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart