SY3H3D67E67E Synthonix Corporation

(9H-Fluoren-9-yl)methyl piperazine-1-carboxylate hydrochloride

  • MDL number MFCD00673790
  • Cas Number : 215190-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59466.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59466.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart