SY3H3D67E688 Synthonix Corporation

(S)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one

  • MDL number MFCD13194798
  • Cas Number : 189028-95-3
  • Molecular Weight : 357.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-3-((S)-5-(4-fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one
Inchi Code 1S/C20H20FNO4/c21-16-11-9-15(10-12-16)18(23)7-4-8-19(24)22-17(13-26-20(22)25)14-5-2-1-3-6-14/h1-3,5-6,9-12,17-18,23H,4,7-8,13H2/t17-,18+/m1/s1
Inchi Key AVAZNWOHQJYCEL-MSOLQXFVSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59537.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/59537.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart