SY3H3D67E68A Synthonix Corporation

(R)-(9H-Fluoren-9-yl)methyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate

  • Cas Number : 215178-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59541.html

Safety Information

MSDS https://synthonix.com/59541.html
Availability details for
Quantity
  • Availability
  • Add to Cart