SY3H3D67E68E Synthonix Corporation

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-3-methylbutanoic acid

  • MDL number MFCD02682577
  • Cas Number : 884880-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59546.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59546.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart