SY3H3D67E68F Synthonix Corporation

5-Fluoro-4,7-bis(5-(trimethylstannyl)thiophen-2-yl)benzo[c][1,2,5]thiadiazole

  • MDL number MFCD30489168
  • Cas Number : 1611002-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59554.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59554.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart