SY3H3D67E6AA Synthonix Corporation

4'-Hydroxyvalerophenone

  • MDL number MFCD00009719
  • Cas Number : 2589-71-1
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Pale yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39654.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39654.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart