SY3H3D67E6B3 Synthonix Corporation

2-Hydrazinylthiazole hydrochloride

  • Cas Number : 30216-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39914.html

Safety Information

MSDS https://synthonix.com/39914.html
Availability details for
Quantity
  • Availability
  • Add to Cart