SY3H3D67E6B8 Synthonix Corporation

6-Hydroxy-5-iodopyrimidin-4(3H)-one

  • MDL number MFCD21742320
  • Cas Number : 1429056-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40035.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40035.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart