SY3H3D67E6BC Synthonix Corporation

5-Hydroxy-1H-pyrazole-3-carboxylic acid

  • Cas Number : 89603-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40078.html

Safety Information

MSDS https://synthonix.com/40078.html
Availability details for
Quantity
  • Availability
  • Add to Cart