SY3H3D67E6C0 Synthonix Corporation

2-Hydroxy-2-methylpropanamide

  • Cas Number : 13027-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40266.html

Safety Information

MSDS https://synthonix.com/40266.html
Availability details for
Quantity
  • Availability
  • Add to Cart