SY3H3D67E6C3 Synthonix Corporation

5-Hydroxypyrimidin-4(3H)-one hydrochloride

  • Cas Number : 1616526-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40320.html

Safety Information

MSDS https://synthonix.com/40320.html
Availability details for
Quantity
  • Availability
  • Add to Cart