SY3H3D67E6C7 Synthonix Corporation

(3S,5S)-5-(Hydroxymethyl)tetrahydrofuran-3-ol

  • MDL number MFCD11226861
  • Cas Number : 204509-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40394.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40394.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart