SY3H3D67E6C9 Synthonix Corporation

(4R,5S)-5-(Hydroxymethyl)tetrahydrofuran-2,4-diol

  • MDL number MFCD00132941
  • Cas Number : 18546-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40397.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40397.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart