SY3H3D67E6D5 Synthonix Corporation

6-Hydrazinyl-9H-purine

  • Cas Number : 5404-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41109.html

Safety Information

MSDS https://synthonix.com/41109.html
Availability details for
Quantity
  • Availability
  • Add to Cart