SY3H3D67E6DA Synthonix Corporation

(5-(Hydroxymethyl)thiophen-3-yl)boronic acid

  • MDL number MFCD27665427
  • Cas Number : 1268683-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41183.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41183.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart