SY3H3D67E701 Synthonix Corporation

2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine

  • Cas Number : 887707-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42026.html

Safety Information

MSDS https://synthonix.com/42026.html
Availability details for
Quantity
  • Availability
  • Add to Cart