SY3H3D67E707 Synthonix Corporation

N-Hydroxypicolinimidoyl chloride

  • MDL number MFCD20257786
  • Cas Number : 69716-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42458.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42458.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart