SY3H3D67E716 Synthonix Corporation

5-Hydroxy-2-methoxylpyridine

  • MDL number MFCD06858737
  • Cas Number : 51834-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to brown solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43140.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43140.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart