SY3H3D67E717 Synthonix Corporation

3-Hydroxy-1-methylpyridin-4(1H)-one

  • MDL number MFCD12405601
  • Cas Number : 50700-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43144.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43144.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart