SY3H3D67E71A Synthonix Corporation

4-Hydroxybenzenesulfonamide

  • Cas Number : 1576-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43166.html

Safety Information

MSDS https://synthonix.com/43166.html
Availability details for
Quantity
  • Availability
  • Add to Cart