SY3H3D67E71C Synthonix Corporation

1-(2-Hydroxyethyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD01011723
  • Cas Number : 936-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43245.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43245.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart