SY3H3D67F2DC Synthonix Corporation

4-Hydroxycyclohexanecarbonitrile

  • MDL number MFCD13688990
  • Cas Number : 24056-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Colorless to Pale-yellow Liquid or Sticky Oil to Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43771.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43771.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart