SY3H3D67F2E1 Synthonix Corporation

N-(4-Hydroxybutyl)acrylamide

  • MDL number MFCD29919236
  • Cas Number : 44915-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystal
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43842.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43842.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart