SY3H3D67F2F5 Synthonix Corporation

5-Hydroxy-2-nitrobenzoic acid

  • MDL number MFCD00017566
  • Cas Number : 610-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45027.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45027.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart